Learn More
Thrombospondin (TSP) Ab-4 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results Epredia™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody.
Marke: Epredia MS-421-B1
Weitere Details : Netto-Gewicht : 0.00190kg
Beschreibung
Thrombospondin is a protein from platelet a-granules. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.Host Species: Mouse
Clone: A6.1
Isotype: IgG1
Species Reactivity: Human, Cow, Pig, Horse, Dog, Sheep, Mouse and Rat. Others not known.
Epitope: Collagen Type V-binding domain of TSP
Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)
Positive Control: Tonsil or Placenta
Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
- Electron Microscopy
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Spezifikation
Thrombospondin (TSP) Ab-4 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Angiogenesis | |
Bovine, Canine, Equine, Human, Mouse, Porcine, Rat, Sheep | |
IgG1 |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
A6.1 | |
Biotin | |
Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets | |
RUO | |
Primary | |
Concentrated |