Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-ubiquitin-FK2 Antibody Kit
LanthaScreen™ Tb-anti-ubiquitin-FK2 Antibody Kit
1058.00 € - 13270.00 €
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
10419603
|
Thermo Scientific™
PV4752 |
25 μg |
1058.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
10504424
|
Thermo Scientific™
PV4751 |
1 mg |
13270.00 €
1 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The Tb-anti-ubiquitin-FK2 antibody preferentially binds to mono- or poly-ubiquitinated proteins over free ubiquitin. This antibody can be used in combination with purified GFP-substrates, fluorescein-ubiquitin, or LanthaScreen™ cell lines to develop robust high-throughput biochemical or cell based assays for ubiquitination or deubiquitination.For Research Use Only. Not for use in diagnostic procedures.