missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCTL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24589-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of family 1 glycosidases. Glycosidases are enzymes that hydrolyze glycosidic bonds and are classified into families based on primary amino acid sequence. Most members of family 1 have two conserved glutamic acid residues, which are required for enzymatic activity. The mouse ortholog of this protein has been characterized and has a domain structure of an N-terminal signal peptide, glycosidase domain, transmembrane domain, and a short cytoplasmic tail. It lacks one of the conserved glutamic acid residues important for catalysis, and its function remains to be determined. Alternative splicing results in multiple transcript variants.Spezifikation
LCTL | |
Polyclonal | |
Unconjugated | |
LCTL | |
KLG, KLPH, lactase like, Lactase like protein, LCTL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
197021 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6UWM7 | |
LCTL | |
LCTL Fusion Protein Ag20220 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |