Learn More
Liothyronine sodium, MedChemExpress
Beschreibung
Liothyronine sodium is an active form of thyroid hormone. Liothyronine sodium is a potent thyroid hormone receptors TRα and TRβ agonist with Kis of 2.33 nM for hTRα and hTRβ, respectively.
Spezifikation
Spezifikation
| Chemischer Name oder Material | Liothyronine sodium |
| CAS | 55-06-1 |
| Farbe | Off-White |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C15H11I3NNaO4 |
| Menge | 100 mg |
| Synonym | Triiodothyronine sodium3,3',5-Triiodo-L-thyronine sodiumT3 sodium |
| Löslichkeitsinformationen | 0.1 M NaOH : 12.5 mg/mL (18.57 mM; ultrasonic and adjust pH to 11 with NaOH) ∣DMSO : 5.56 mg/mL (8.26 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | N[C@@H](CC1=CC(I)=C(C(I)=C1)OC2=CC=C(O)C(I)=C2)C(O[Na])=O |
| Molekulargewicht (g/mol) | 672.96 |
| Mehr anzeigen |