missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY341495, MedChemExpress
Beschreibung
LY341495 is a metabotropic glutamate receptor (mGluR) antagonist with IC50s of 21 nM, 14 nM, 7.8 μM, 8.2 μM, 170 nM, 990 nM, 22 μM for mGlu2, mGlu3, mGlu1a, mGlu5a, mGlu8, mGlu7, and mGlu4 receptors, respectively.
Spezifikation
Spezifikation
| Chemischer Name oder Material | LY341495 |
| CAS | 201943-63-7 |
| Farbe | Light Yellow |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C20H19NO5 |
| Menge | 25 mg |
| Löslichkeitsinformationen | DMSO : 6 mg/mL (16.98 mM; Need ultrasonic) |
| SMILES | O=C(O)[C@@H]1[C@H](C1)[C@@](N)(CC2C3=C(C=CC=C3)OC4=C2C=CC=C4)C(O)=O |
| Molekulargewicht (g/mol) | 353.37 |
| Formelmasse | 353.37 |
| Mehr anzeigen |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?