missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl N-(3,5-dichlorphenyl)carbamat, Thermo Scientific™
Click to view available options
Menge:
10 g
25 g
50 g
Spezifikation
Chemischer Name oder Material | Methyl N-(3, 5-dichlorophenyl)carbamate |
CAS Min. % | 97.0 |
Prozentgehaltsbereich | 97% |
Menge | 10 g |
Formelmasse | 220.05 |
Reinheit (%) | 97% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'