Learn More
ACROBiosystems Mouse IL-15 Protein, Fc Tag
Marke: ACROBiosystems IL5-M5255-50ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Interleukin 15 is also known as IL15, IL-15, and is a cytokine with structural similarity to IL-2. Like IL-2, IL-15 binds to and signals through the IL-2/IL-15 beta chain (CD122) and the common gamma chain (gamma-C, CD132). IL-15 is secreted by mononuclear phagocytes (and some other cells) following infection by virus(es). This cytokine induces cell proliferation of natural killer cells; cells of the innate immune system whose principal role is to kill virally infected cells. Interleukin 15 (IL-15) regulates T and natural killer (NK) cell activation and proliferation. Survival signals that maintain memory T cells in the absence of antigen are provided by IL-15. This cytokine is also implicated in NK cell development. In rodent lymphocytes, IL-15 prevents apoptosis by inducing an apoptosis inhibitor, BCL2L1/BCL-x(L). IL-15 has been shown to enhance the anti-tumor immunity of CD8+ T cells in pre-clinical models. A phase I clinical trial to evaluate the safety, dosing, and anti-tumor efficacy of IL-15 in patients with metastatic melanoma and renal cell carcinoma (kidney cancer) has begun to enroll patients at the National Institutes of Health.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 49 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_032383.1 | |
50 μg | |
Asn 49 - Ser 162 | |
IL-15,Interleukin-15,MGC9721 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
39.7 kDa | |
Mouse IL-15, Fc Tag (IL5-M5255) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P48346-1). | |
-20°C | |
IL-15 | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Cytokines and Growth Factors |