Learn More
ACROBiosystems Mouse IL-15 Protein, Fc Tag
423.38 € - 3495.90 €
Spécification
Zugriffsnummer | NP_032383.1 |
---|---|
Molekulargewicht | 39.7 kDa |
Quelle | Mouse IL-15, Fc Tag (IL5-M5255) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P48346-1). |
Immunogen | Asn 49 - Ser 162 |
Lagerungsbedingungen | -20°C |
Code produit | Marque | Menge | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Menge | Prix | Quantité et disponibilité | |||||
16467709
|
ACROBiosystems
IL5-M5255-50UG |
50 μg |
423.38 €
50 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16457709
|
ACROBiosystems
IL5-M5255-1MG |
1 mg |
3495.90 €
1 Milligramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Description
Interleukin 15 is also known as IL15, IL-15, and is a cytokine with structural similarity to IL-2. Like IL-2, IL-15 binds to and signals through the IL-2/IL-15 beta chain (CD122) and the common gamma chain (gamma-C, CD132). IL-15 is secreted by mononuclear phagocytes (and some other cells) following infection by virus(es). This cytokine induces cell proliferation of natural killer cells; cells of the innate immune system whose principal role is to kill virally infected cells. Interleukin 15 (IL-15) regulates T and natural killer (NK) cell activation and proliferation. Survival signals that maintain memory T cells in the absence of antigen are provided by IL-15. This cytokine is also implicated in NK cell development. In rodent lymphocytes, IL-15 prevents apoptosis by inducing an apoptosis inhibitor, BCL2L1/BCL-x(L). IL-15 has been shown to enhance the anti-tumor immunity of CD8+ T cells in pre-clinical models. A phase I clinical trial to evaluate the safety, dosing, and anti-tumor efficacy of IL-15 in patients with metastatic melanoma and renal cell carcinoma (kidney cancer) has begun to enroll patients at the National Institutes of Health.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 49 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_032383.1 | |
Mouse IL-15, Fc Tag (IL5-M5255) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P48346-1). | |
-20°C | |
IL-15 | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Cytokines and Growth Factors |
39.7 kDa | |
Asn 49 - Ser 162 | |
IL-15,Interleukin-15,MGC9721 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |