Learn More
ACROBiosystems Mouse Nectin-4 Protein, His Tag
Marke: ACROBiosystems NE4-M52H3-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Nectin-4 (gene name PVRL4, poliovirus receptor-like 4) is a 66 kDa type I transmembrane glycoprotein belonging to the Nectin family of Ig superfamily proteins. Nectins are cell adhesion molecules that play a key role in various biological processes such as polarity, proliferation, differentiation and migration, for epithelial, endothelial, immune and neuronal cells, during development and adult life. Nectin-4 is a tumor-associated antigen in 50%, 49% and 86%o of breast, ovarian and lung carcinomas, respectively, mostly on tumors of bad prognosis. Its expression is not detected in the corresponding normal tissues.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.2 kDa. The protein migrates as 38-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_082169.2 | |
1 mg | |
Gly 31 - Ser 347 | |
NECTIN4,LNIR, PRR4, PVRL4 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
36.2 kDa | |
Mouse Nectin-4, His Tag (NE4-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 31 - Ser 347 (Accession # Q8R007-1). | |
-20°C | |
Nectin-4 | |
Mouse | |
PBS, Arginine, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |