missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse Nectin-4 Protein, His Tag
423.38 € - 3108.82 €
Spezifikation
Zugriffsnummer | NP_082169.2 |
---|---|
Molekulargewicht | 36.2 kDa |
Quelle | Mouse Nectin-4, His Tag (NE4-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 31 - Ser 347 (Accession # Q8R007-1). |
Immunogen | Gly 31 - Ser 347 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16497909
|
ACROBiosystems
NE4-M52H3-100UG |
100 μg |
423.38 €
100 Mikrogramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16407919
|
ACROBiosystems
NE4-M52H3-1MG |
1 mg |
3108.82 €
1 Milligramm |
Verfügbar ab: 22-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Nectin-4 (gene name PVRL4, poliovirus receptor-like 4) is a 66 kDa type I transmembrane glycoprotein belonging to the Nectin family of Ig superfamily proteins. Nectins are cell adhesion molecules that play a key role in various biological processes such as polarity, proliferation, differentiation and migration, for epithelial, endothelial, immune and neuronal cells, during development and adult life. Nectin-4 is a tumor-associated antigen in 50%, 49% and 86%o of breast, ovarian and lung carcinomas, respectively, mostly on tumors of bad prognosis. Its expression is not detected in the corresponding normal tissues.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.2 kDa. The protein migrates as 38-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_082169.2 | |
Mouse Nectin-4, His Tag (NE4-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 31 - Ser 347 (Accession # Q8R007-1). | |
-20°C | |
Nectin-4 | |
Mouse | |
PBS, Arginine, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
36.2 kDa | |
Gly 31 - Ser 347 | |
NECTIN4,LNIR, PRR4, PVRL4 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |