missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRE11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10744-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Mre11 is an 80 kDa protein that acts as a Mn+2 dependent double stranded DNA 3′ to 5′ exonuclease and a single stranded DNA endonuclease. It is part of a larger complex with Rad50, p95 (Nibrin) and possibly other associated proteins involved in the nucleolytic end-processing of double strand breaks and telomere maintece.Spezifikation
MRE11 | |
Polyclonal | |
Unconjugated | |
MRE11 | |
ATLD, HNGS1, MRE11, MRE11 homolog 1, MRE11 homolog A, MRE11A, MRE11B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17535, 4361, 64046 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49959, Q61216 | |
MRE11, Mre11a | |
MRE11 Fusion Protein Ag1105 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |