Learn More
N-(2,6-Difluorphenyl)-4-Methoxybenzamid, 97 %, Thermo Scientific™
Catalog Number 15465376
Click to view available options
Menge:
250 mg
1 g
155997-19-6
C14H11F2NO2
263.24
MFCD04310051
NDUMEFRGYXBRDG-UHFFFAOYSA-N
n-2,6-difluorophenyl-4-methoxybenzamide, chembl45855, n-2,6-difluoro-phenyl-4-methoxy-benzamide, n-2,6-difluorophenyl 4-methoxyphenyl carboxamide
956606
N-(2,6-difluorphenyl)-4-methoxybenzamid
COC1=CC=C(C=C1)C(=O)NC1=C(F)C=CC=C1F
155997-19-6
C14H11F2NO2
263.24
MFCD04310051
NDUMEFRGYXBRDG-UHFFFAOYSA-N
n-2,6-difluorophenyl-4-methoxybenzamide, chembl45855, n-2,6-difluoro-phenyl-4-methoxy-benzamide, n-2,6-difluorophenyl 4-methoxyphenyl carboxamide
956606
N-(2,6-difluorphenyl)-4-methoxybenzamid
COC1=CC=C(C=C1)C(=O)NC1=C(F)C=CC=C1F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-(2,6-Difluorophenyl)-4-methoxybenzamide
Menge 250 mg
Formelmasse 263.24
Reinheit (%) 97%