missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1-(p-Toluolsulfonyl)-L-Histidin-Dicyclohexylammoniumsalz, 98 %, Thermo Scientific™

Catalog Number 15467156
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
65057-34-3
C30H46N4O6S
590.78
MFCD00039005
RNRUVXUHYUOWJW-RSAXXLAASA-N
boc-his tos-oh.dcha, dicyclohexylamine s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoate, boc-his toc-oh.dcha, n-tert-butoxycarbonyl-1-p-tosyl-l-histidine, compound with dicyclohexylamine 1:1, n-alpha-t-butyloxycarbonyl-n-im-t-tosyl-l-histidine dicyclohexylamine, boc-his toc-oh?cha, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid; dicha, l-histidine, n-1,1-dimethylethoxy carbonyl-1-4-methylphenyl sulfonyl-, compd with n-cyclohexylcyclohexanamine 1:1, n-cyclohexylcyclohexanamine; 2s-3-1-4-methylphenyl sulfonylimidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid
6455009
C1CCC(CC1)NC1CCCCC1.CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
65057-34-3
C30H46N4O6S
590.78
MFCD00039005
RNRUVXUHYUOWJW-RSAXXLAASA-N
boc-his tos-oh.dcha, dicyclohexylamine s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoate, boc-his toc-oh.dcha, n-tert-butoxycarbonyl-1-p-tosyl-l-histidine, compound with dicyclohexylamine 1:1, n-alpha-t-butyloxycarbonyl-n-im-t-tosyl-l-histidine dicyclohexylamine, boc-his toc-oh?cha, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid; dicha, l-histidine, n-1,1-dimethylethoxy carbonyl-1-4-methylphenyl sulfonyl-, compd with n-cyclohexylcyclohexanamine 1:1, n-cyclohexylcyclohexanamine; 2s-3-1-4-methylphenyl sulfonylimidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid
6455009
C1CCC(CC1)NC1CCCCC1.CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-1-(p-toluenesulfonyl)-L-histidine dicyclohexylammonium salt
IUPAC-Name N-Cyclohexylcyclohexanamin;(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propansäure
Formelmasse 590.77
Reinheit (%) 98%
Menge 5 g
Physikalische Form Fest
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'