missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15604-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
NAPG is soluble NSF attachment protein gamma. The soluble NSF attachment proteins (SNAPs) enable N-ethyl-maleimide-sensitive fusion protein (NSF) to bind to target membranes. NSF and SNAPs appear to be general components of the intracellular membrane fusion apparatus, and their action at specific sites of fusion must be controlled by SNAP receptors particular to the membranes being fused. The product of this gene mediates platelet exocytosis and controls the membrane fusion events of this process.Spezifikation
NAPG | |
Polyclonal | |
Unconjugated | |
NAPG | |
GAMMASNAP, NAPG, SNAP gamma, SNAPG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108123, 307382, 8774 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99747, Q9CWZ7 | |
NAPG | |
NAPG Fusion Protein Ag7988 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |