missing translation for 'onlineSavingsMsg'
Learn More

Neuropilin-1 (CD304) Mouse anti-Human, Alexa Fluor 647, Clone: U21-1283, BD Pharmingen™

Mouse Monoclonal Antibody

Marke:  BD Biosciences 566048

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15884709

  • 265.00 € / 1 Stück
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.

Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.
Spezifikation

Spezifikation

Neuropilin-1 (CD304)
Monoclonal
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
25 Tests
Cell Biology
Human
Purified
Flow Cytometry
U21-1283
Vol. Per test: 5μL
Neuropilin; NRP; NP1; Blood dendritic cell antigen 4; BDCA-4; VEGF165R
Recombinant Human Neuropilin-1
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed.
IgG2b κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.