missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p57Kip2 Mouse anti-Human, Mouse, Rat, Clone: 1A12H3, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66794-1-IG-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.Spezifikation
p57Kip2 | |
Monoclonal | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49918, P49919 | |
CDKN1C | |
p57Kip2 Fusion Protein Ag19927 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG1 |
Western Blot | |
1A12H3 | |
Unconjugated | |
CDKN1C | |
BWCR, BWS, CDKN1C, KIP2, p57, p57 Kip2, p57Kip2, WBS | |
Mouse | |
Protein G | |
RUO | |
1028, 12577, 246060 | |
-20°C | |
Liquid |