Learn More
DiscoverX PathHunter™ eXpress KISS1R U2OS β-Arrestin GPCR Assay
Beschreibung
The PathHunter™ KISS1R β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. kisspeptin) based activation of KISS1R GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged KISS1R and EA-tagged β-Arrestin 2. Activation of KISS1R stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Spezifikation
Spezifikation
| Assay-Eingabe | β-Arrestin-2 recruitment |
| Zur Verwendung mit (Anwendung) | Assay-Ready eXpress Kits |
| Primärer Agonist | Kisspeptin-10 |
| Menge | 1.000 dp (10 x 96-Well) |
| Zieleintrag | KISS1R |
| Zelllinie | U2OS |
| Ziel | KISS1R |