missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ PC Cryogenic Vials Racks
Marke: Carl Roth™ K554.1
Weitere Details : Netto-Gewicht : 0.25000kg
Specifications
Violet | |
200 mm | |
200 x 100 x 25 mm | |
1 Pack | |
Polypropylene |
25 mm | |
100 mm | |
Used as Stands For Cryo Tubes | |
200 x 100 mm | |
Cryogenic Vial Rack |