Learn More
PD98059, MedChemExpress
Beschreibung
PD98059 binds to the inactive form of MEK, thereby preventing the activation of MEK1 (IC50 of 2-7 μM) and MEK2 (IC50 of 50 μM) by upstream kinases. PD98059 is a ERK1/2 signaling inhibitor. PD98059 is a ligand for the aryl hydrocarbon receptor (AHR), and suppresses TCDD binding (IC50 of 4 μM) and AHR transformation (IC50 of 1 μM). PD98059 also inhibits autophagy.
Spezifikation
Spezifikation
| Chemischer Name oder Material | PD98059 |
| CAS | 167869-21-8 |
| Farbe | Yellow |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C16H13NO3 |
| Menge | 100 mg |
| Löslichkeitsinformationen | DMSO : 33.33 mg/mL (124.70 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1C=C(OC2=CC=CC=C21)C3=CC=CC(OC)=C3N |
| Molekulargewicht (g/mol) | 267.28 |
| Formelmasse | 267.28 |
| Mehr anzeigen |