missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-AKT400, MedChemExpress
Description
PF-AKT400 is a broadly selective, potent, ATP-competitive Akt inhibitor, displays 900-fold greater selectivity for PKBα (IC50=0.5 nM) than PKA (IC50=450 nM).
Specifications
Specifications
| Chemischer Name oder Material | PF-AKT400 |
| CAS | 1004990-28-6 |
| Farbe | White |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C20H22F2N6O |
| Menge | 10 mg |
| Synonym | AKT protein kinase inhibitor |
| Löslichkeitsinformationen | DMSO : ≥ 100 mg/mL (249.73 mM) |
| SMILES | N[C@@](CC1)(CNC(C2=CC=C(F)C=C2F)=O)CN1C3=C4C(NC=C4CC)=NC=N3 |
| Molekulargewicht (g/mol) | 400.43 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?