missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTK2B (FAK2) Recombinant Human Protein
PTK2B (FAK2) is a cytoplasmic tyrosine kinase.
Marke: Invitrogen™ PR8161B
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PTK2B (FAK2) is involved in calcium-induced regulation of ion channels and activation of the map kinase signaling pathway.
Spezifikation
NP_004094 | |
50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
PTK2B (FAK2) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
145.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
FAK Kinase Family | |
Recombinant | |
Baculovirus |