missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ F-Actin Labeling Reagents

Artikelnummer. 32214192
Ansicht ändern
Click to view available options
Farbstofftyp:
Alexa Fluor™ 488
Alexa Fluor™ 568
Alexa Fluor™ Plus 647
TRITC (Tetramethylrhodaminisothiocyanat)
Anregungswellenlängenbereich:
495/518
540/565
580/603
658/675
Packungsgröße:
1 Stück
5 Milliliter
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Farbstofftyp Anregungswellenlängenbereich Packungsgröße
32214192 Alexa Fluor™ Plus 647 658/675 5 Milliliter
14879680 Alexa Fluor™ 488 495/518 5 Milliliter
15119325 TRITC (Tetramethylrhodaminisothiocyanat) 540/565 1 Stück
31935074 Alexa Fluor™ 568 580/603 5 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 32214192 Lieferant Invitrogen™ Lieferanten-Nr. R37179
 Eingeschränkter Artikel
Verfügbar ab: 16-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ActinRed™ and ActinGreen™ ReadyProbes™ reagents assist with bright, selective F-actin labeling in fixed cells and tissues. The ready-to-use dropper or dual-use bottle enables dropwise dispensing or direct pipetting for fast, flexible cytoskeleton imaging.

ActinGreen 488 ReadyProbes Reagent is a selective, high-affinity F-actin probe conjugated to our bright, photostable, green-fluorescent Alexa Fluor 488 dye. ActinGreen 488 reagent is a room temperature-stable solution of Alexa Fluor 488 ActinGreen (Cat. No. A12379) packaged in a dropper bottle. ActinRed 555 ReadyProbes Reagent is a comparable F-actin stain conjugated to the red-orange fluorescent dye tetramethylrhodamine (TRITC). ActinRed 555 reagent is a room temperature-stable solution of Rhodamine F-actin probe (Cat. No. R415) packaged in a dropper bottle.

ActinRed 568 ReadyProbes Reagent is our high-affinity F-actin probe conjugated to our bright, red-fluorescent Alexa Fluor 568 dye. This dye is packaged in an easy to use dropper-optional bottle, allowing end-users to choose either to pipet out of the tube using the classic cap, or utilize the dropper bottle tip for typical ReadyProbes workflows.

ActinRed 647 ReadyProbes Reagent leverages our high-affinity F-actin probe conjugated to our bright Far-Red Alexa Fluor 647 Plus dye, offering enhanced brightness vs. classic Alexa Fluor 647 F-actin probe conjugates, in an easy to use format. ActinRed 647 ReadyProbes Reagent should be stored from 2-8°C for long-term, and also offers the option to leverage pipetting, if desired, via the dropper-optional cap.

These ready-to-use solutions combine superior brightness with exceptional F-actin staining. ReadyProbes actin has several advantages over antibodies for actin labeling, including virtually identical binding properties with actin from different species of plants and animals and low non-specific binding.

TRUSTED_SUSTAINABILITY

Spezifikation

Farbe Far Red
Konzentration Ready-to-use
Inhalt und Lagerung •1 Dual-use Bottle
•Store at 15°C to 30°C
•Protect from light
•Do not freeze
Anregungswellenlängenbereich 658/675
Farbstofftyp Alexa Fluor™ Plus 647
Format Dual-use Dropper Bottle
Zur Verwendung mit (Anwendung) Fixed Cell Imaging, High Content Screening, Fluorescence Microscopy
Zur Verwendung mit (Geräte) Confocal Microscope, EVOS™, CellInsight™ Cell Imaging Instruments, Fluorescence Microscope, Flow Cytometer
Anzahl Tests 75 Tests
Enthält Dropper Tip, Dropper Cap
Reaktivität F-actin
Menge 1 Dual-use bottle
Volumen (metrisch) 6 mL
Nachweisverfahren Fluorescence
Liefertyp Add 2 drops (80 μL) per 1 mL of medium
Emission 675 nm
Form Ready-to-use aqueous solution
Produktlinie ActinRed, ReadyProbes
Ziel F-actin Cytoskeleton
Verfahren Cell Paint Assay
Markertyp Fluorescent Dye
Produkttyp Cell Labeling
Sub Cellular Localization Cytoskeleton
Zellpermeabilität Fixed Cells
Synonym Phalloidin
Anregung 658 nm
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.