missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Leptin Receptor Protein
A cDNA sequence encoding the Leptin Receptor was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12554-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Leptin Receptor Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein was filter sterilized and stored at 4C (0.2 to 0.5 mg/ml) solution of Tris-HCl buffer, pH 9.0 with 150mM NaCl. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chicken | |
Untagged | |
Greater than 99.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |