missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ebola Virus Ebola Zaire Protein
A cDNA sequence encoding the Ebola Zaire Protein was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11728-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Specifications
Ebola Zaire Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Ebola Zaire protein solution is supplied in Phosphate buffer and 0.02% sodium azide. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Ebola Virus | |
His | |
>95% pure as determined by 12% SDS-PAGE (Coomassie blue stain). |