Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5B Protein
A cDNA sequence encoding the HCV NS5B was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12200-500ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Hepatitis C HCV NS5B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
His | |
Protein is >95% pure as determined by SDS-PAGE. |