missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV HTLV-1 p24 core Protein
A cDNA sequence encoding the HTLV-1 p24 core was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12358-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
HIV HTLV-1 p24 core Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM NaPO4 pH 6.0, containing 1mM DTT and 1mM EDTA. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
HTLV-1 p24 protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |