missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BEND6 Protein
A cDNA sequence encoding the BEND6 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11153-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
BEND6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQKIVQT DEITNTQAFR KGKRKRTETM DSENANSDMD KGQRDPYSGN AFLPGESSSE DEEPLAELSK EELCAKIKSL KEKLTNTRKE NSRLRQSLVM LQVLPQAVTQ FEELVGMAEA LLKGGGTMST SASTLWRATN NSSPDSFAST CSNSNSNSSS PVSLKPEEEH QTDEKQFQIE KWQIARCNKS KPQKFINDLM QVLYTNEYMA THSLTGAKSS TSRDKAVKPA MNQNEVQEII GVTKQLFPNT DDVSIRRMIG QKLNNCTKKP NLSKNLNSQD IK | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BEND6 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |