missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BMPR1B Protein
A cDNA sequence encoding the BMPR1B was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11173-2ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
658 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSKKEDGES TAPTPRPKVL RCKCHHHCPE DSVNNICSTD GYCFTMIEED DSGLPVVTSG CLGLEGSDFQ CRDTPIPHQR RSIECCTERN ECNKDLHPTL PPLKNRDFVD GPIHHR | |
Greater than 80.0% as determined by SDS-PAGE. |
BMPR1B Protein | |
Research Use Only | |
BMPR1B | |
Human | |
His | |
BMPR1B protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol, 1mM DTT and 0.1mM PMSF. |