missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNKSR2 Protein
A DNA sequence encoding the Homo sapiens (Human) CNKSR2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP9073-ye-200ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_001162118.1 | |
CNKSR2 Protein | |
Research Use Only | |
CNKSR2 | |
Recombinant Protein | |
Yeast | |
AAEHLDDMNR WLNRINMLTA GYAERERIKQ EQDYWSESDK EEADTPSTPK QDSPPPPYDT YPRPPSMSCA SPYVEAKHSR LSSTETSQSQ SSHEEFRQEV TGSSAVSPIR KTASQRRSWQ DLIETPLTSS GLHYLQTLPL EDSVFSDSAA I | |
Greater than 90% as determined by SDS-PAGE. |
22866 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |