missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTRB1 Protein
A cDNA sequence encoding the CTRB1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10565-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
CTRB1 Protein | |
Research Use Only | |
1100 units/mg protein. One unit is defined as the amount of enzyme that will hydrolyze 1.0 mole of N-alpha-acetyl-L-tyrosine ethyl ester (ATEE) per min at pH 7.0 at 25C. | |
Human | |
Untagged | |
The Human CTRB1 was lyophilized without any additives. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
CG VPAIHPVLSG LSRIVNGEDA VPGSWPWQVS LQDKTGFHFC GGSLISEDWV VTAAHCGVRT SDVVVAGEFD QGSDEENIQV LKIAKVFKNP KFSILTVNND ITLLKLATPA RFSQTVSAVC LPSADDDFPAGTLCATTGWG KTKYNANKTP DKLQQAALPL LSNAECKKSW GRRITDVMIC AGASGVSSCM GDSGGPLVCQ KDGAWTLVGI VSWGSDTCST SSPGVYARVTKLIPWVQKIL AAN | |
Greater than 95% as determined by HPLC. |