Learn More
R&D Systems™ Recombinant Human CX3CL1/Fractalkine (Full Length) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke:  R&D Systems™ 365-FR-025/CF
Weitere Details : Netto-Gewicht : 0.00970kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 18345007
  • 485.95 € / 25 Mikrogramm
Verfügbar ab: 23-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
The Recombinant Human CX3CL1/Fractalkine (Full Length) is derived from NS0. The Recombinant Human CX3CL1/Fractalkine (Full Length) has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS.
35 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
6376
CX3CL1/Fractalkine Full Length
Mouse myeloma cell line,NS0-derived human CX3CL1/Fractalkine protein, (N-terminus) Human CX3CL 1 (Gln25-Arg339) (Ser199Asn) Accession &Num; P78423 KQN 6-His tag (C-terminus)
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.