missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM3B Protein
A cDNA sequence encoding the FAM3B was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11845-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
FAM3B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS ELIPDAPLSS AAYSIRSIGE RPVLKAPVPK RQKCDHWTPC PSDTYAYRLL SGGGRSKYAK ICFEDNLLMG EQLGNVARGI NIAIVNYVTG NVTATRCFDM YEGDNSGPMT KFIQSAAPKS LLFMVTYDDG STRLNNDAKN AIEALGSKEI RNMKFRSSWV FIAAKGLELP SEIQREKINH SDAKNNRYSG WPAEIQIEGC IPKERS | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FAM3B Human was filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 20mM TRIS, 50mM NaCl, pH 7.5 |