missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-F Protein
A cDNA sequence encoding the HLA-F was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12277-10ug
Additional Details : Netto-Gewicht : 0.01000kg
Specifications
3134 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGSHSLRY FSTAVSRPGR GEPRYIAVEY VDDTQFLRFD SDAAIPRMEP REPWVEQEGP QYWEWTTGYA KANAQTDRVA LRNLLRRYNQ SEAGSHTLQG MNGCDMGPDG RLLRGYHQHA YDGKDYISLN EDLRSWTAAD TVAQITQRFY EAEEYAEEFR TYLEGECLEL LRRYLENGKE TLQRADPPKA HVAHHPISDH EATLRCWALG FYPAEITLTW QRDGEEQTQD TELVETRPAG DGTFQKWAAV VVPPGEEQRY TCHVQHEGLP QPLILRWEQS PQPTIPI | |
Greater than 90.0% as determined by SDS-PAGE. |
HLA-F Protein | |
Research Use Only | |
HLA-F | |
Human | |
His | |
HLA-F protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl 20% glycerol and 1mM DTT. |