missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSPB8 Protein
A cDNA sequence encoding the HSPB8 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12335-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
26353 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MADGQMPFSC HYPSRLRRDP FRDSPLSSRL LDDGFGMDPF PDDLTASWPD WALPRLSSAW PGTLRSGMVP RGPTATARFG VPAEGRTPPP FPGEPWKVCV NVHSFKPEEL MVKTKDGYVE VSGKHEEKQQ EGGIVSKNFT KKIQLPAEVD PVTVFASLSP EGLLIIEAPQ VPPYSTFGES SFNNELPQDS QEVTCT | |
Greater than 95.0% as determined by SDS-PAGE. |
HSPB8 Protein | |
Research Use Only | |
HSPB8 | |
Human | |
Untagged | |
The HSPB8 protein was lyophilized from a concentrated (1 mg/ml) solution containing 20mM Tris-acetate, pH 7.6, 10mM NaCl, 0.1mM EDTA, 0.1mM PMSF, 15mM b-ME. |