missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF1 Isoform 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Interferon alpha 10 / IFNA10, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP5005-20 ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_002162.1 | |
IGF1 Isoform 2 Protein | |
Research Use Only | |
IFNA10 | |
Recombinant Protein | |
Yeast | |
CDLPQTHSLG NRRALILLGQ MGRISPFSCL KDRHDFRIPQ EEFDGNQFQK AQAISVLHEM IQQTFNLFST EDSSAAWEQS LLEKFSTELY QQLNDLEACV IQEVGVEETP LMNEDSILAV RKYFQRITLY LIERKYSPCA WEVVRAEIMR SLSFSTNLQK RLRRKD | |
Greater than 90% as determined by SDS-PAGE. |
3446 | |
20 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |