missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1RL1 Protein
A cDNA sequence encoding the IL1RL1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12413-HIS-EC-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
9173 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGFWILAILT ILMYSTAAKF SKQSWGLENE ALIVRCPRQG KPSYTVDWYY SQTNKSIPTQ ERNRVFASGQ LLKFLPAAVA DSGIYTCIVR SPTFNRTGYA NVTIYKKQSD CNVPDYLMYS TVSGSEKNSK IYCPTIDLYN WTAPLEWFKN CQALQGSRYR AHKSFLVIDN VMTEDAGDYT CKFIHNENGA NYSVTATRSF TVKDEQGFSL FPVIGAPAQN EIKEVEIGKN ANLTCSACFG KGTQFLAAVL WQLNGTKITD FGEPRIQQEE GQNQSFSNGL ACLDMVLRIA DVKEEDLLLQ YDCLALNLHG LRRHTVRLSR KNPIDHHSIY CIIAVCSVFL MLINVLVIIL KMFWIEATLL WRDIAKPYKT RNDGKLYDAY VVYPRNYKSS TDGASRVEHF VHQILPDVLE NKCGYTLCIY GRDMLPGEDV VTAVETNIRK SRRHIFILTP QITHNKEFAY EQEVALHCAL IQNDAKVILI EMEALSELDM LQAEALQDSL QHLMKVQGTI KWREDHIANK RSLNSKFWKH VRYQMPVPSK IPRKASSLTP LAAQKQ | |
Greater than 95.0% as determined by SDS-PAGE. |
IL1RL1 Protein | |
Research Use Only | |
IL1RL1 | |
Human | |
His | |
IL1RL1 protein solution is supplied in 25mM Sodium Acetate pH 4.8 and 50% glycerol. |