missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IRF 1 Protein
A cDNA sequence encoding the IRF 1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12456-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
3659 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPITRMRMRP WLEMQINSNQ IPGLIWINKE EMIFQIPWKHAAKHGWDINK DACLFRSWAI HTGRYKAGEK EPDPKTWKAN FRCAMNSLPD IEEVKDQSRN KGSSAVRVYR MLPP | |
Greater than 90.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IRF 1 Protein | |
Research Use Only | |
IRF1 | |
Human | |
His | |
1 mg/ml in 20mM Tris pH 8 and 10% glycerol. |