missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KIN Protein
A cDNA sequence encoding the KIN was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12486-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
KIN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGKSDFL TPKAIANRIK SKGLQKLRWY CQMCQKQCRDENGFKCHCMS ESHQRQLLLA SENPQQFMDY FSEEFRNDFL ELLRRRFGTK RVHNNIVYNE YISHREHIHM NATQWETLTD FTKWLGREGL CKVDETPKGW YIQYIDRDPE TIRRQLELEKKKKQDLDDEE KTAKFIEEQV RRGLEGKEQE VPTFTELSRE NDEEKVTFNL SKGACSSSGATSSKSSTLGP SALKTIGSSA SVKRKESSQS STQSKEKKKK KSALDEIMEI EEEKKRTARTDYWLQPEIIV KIITKKLGEK YHKKKAIVKE VIDKYTAVVK MIDSGDKLKL DQTHLETVIPAPGKRILVLN GGYRGNEGTL ESINEKTFSA TIVIETGPLK GRRVEGIQYE DISKLA | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The KIN solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl and 20% glycerol. |