missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLK13 Protein
A cDNA sequence encoding the KLK13 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12503-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
KLK13 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGGVSQES SKVLNTNGTS GFLPGGYTCF PHSQPWQAAL LVQGRLLCGG VLVHPKWVLT AAHCLKEGLK VYLGKHALGR VEAGEQVREV VHSIPHPEYR RSPTHLNHDH DIMLLELQSP VQLTGYIQTL PLSHNNRLTP GTTCRVSGWG TTTSPQVNYP KTLQCANIQL RSDEECRQVY PGKITDNMLC AGTKEGGKDS CEGDSGGPLV CNRTLYGIVS WGDFPCGQPD RPGVYTRVSR YVLWIRETIR KYETQQQKWL KGPQ | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KLK13 protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |