missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LAG-1 Protein
A cDNA sequence encoding the LAG-1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12523-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
388372; 9560 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAPMGS DPPTACCFSY TARKLPRNFV VDYYETSSLC SQPAVVFQTK RGKQVCADPS ESWVQEYVYD LELN | |
Greater than 90.0% as determined by SDS-PAGE. |
LAG-1 Protein | |
Research Use Only | |
CCL4L1; CCL4L2 | |
Human | |
His | |
LAG-1 protein solution (0.5 mg/ml) containing 10mM sodium citrate (pH 3.5) and 10% glycerol. |