missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIN7C Protein
A cDNA sequence encoding the LIN7C was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12575-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
LIN7C Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGCSHHHHHH SSGLVPRGSH MGSMAALGEP VRLERDICRA IELLEKLQRS GEVPPQKLQA LQRVLQSEFC NAVREVYEHV YETVDISSSP EVRANATAKA TVAAFAASEG HSHPRVVELP KTEEGLGFNI MGGKEQNSPI YISRIIPGGI ADRHGGLKRG DQLLSVNGVS VEGEHHEKAV ELLKAAQGKV KLVVRYTPKV LEEMESRFEK MRSAKRRQQT | |
Greater than 90.0% as determined by SDS-PAGE |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LIN7C protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl and 30% glycerol. |