missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LYPLAL1 Protein
A cDNA sequence encoding the LYPLAL1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12607-20ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
127018 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAAASG SVLQRCIVSP AGRHSASLIF LHGSGDSGQG LRMWIKQVLN QDLTFQHIKI IYPTAPPRSY TPMKGGISNV WFDRFKITND CPEHLESIDV MCQVLTDLID EEVKSGIKKN RILIGGFSMG GCMAMHLAYR NHQDVAGVFA LSSFLNKASA VYQALQKSNG VLPELFQCHG TADELVLHSW AEETNSMLKS LGVTTKFHSF PNVYHELSKT ELDILKLWIL TKLPGEMEKQ K | |
Greater than 95% as determined by SDS-PAGE. |
LYPLAL1 Protein | |
Research Use Only | |
LYPLAL1 | |
Human | |
His | |
The LYPLAL1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |