missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MOSPD1 Protein
A cDNA sequence encoding the MOSPD1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12721-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
MOSPD1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMHQQKRQ PELVEGNLPV FVFPTELIFY ADDQSTHKQV LTLYNPYEFA LKFKVLCTTP NKYVVVDAAG AVKPQCCVDI VIRHRDVRSC HYGVIDKFRL QVSEQSQRKA LGRKEVVATL LPSAKEQQKE EEEKRLKEHL TESLFFEQSF QPENRAVSSG P | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MOSPD1 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 0.15M NaCl, 50% glycerol and 1mM DTT. |