missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neurexophilin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Neurexophilin-2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10093-ma-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_009157.1 | |
Neurexophilin-2 Protein | |
Research Use Only | |
NXPH2 | |
Recombinant Protein | |
Mammal | |
KEVVHATEGL DWEDKDAPGT LVGNVVHSRI ISPLRLFVKQ SPVPKPGPMA YADSMENFWD WLANITEIQE PLARTKRRPI VKTGKFKKMF GWGDFHSNIK TVKLNLLITG KIVDHGNGTF SVYFRHNSTG LGNVSVSLVP PSKVVEFEVS PQSTLETKES KSFNCRIEYE KTDRAKKTAL CNFDPSKICY QEQTQSHVSW LCSKPFKVIC IYIAFYSVDY KLVQKVCPDY NYHSETPYLS SG | |
Greater than 90% as determined by SDS-PAGE. |
11249 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |