missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Programmed cell death protein 1
A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 5, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6475-ec-50ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_004699.1 | |
Programmed cell death protein 1 | |
Research Use Only | |
PDCD5 | |
Recombinant Protein | |
E. coli | |
ADEELEALRR QRLAELQAKH GDPGDAAQQE AKHREAEMRN SILAQVLDQS ARARLSNLAL VKPEKTKAVE NYLIQMARYG QLSEKVSEQG LIEILKKVSQ QTEKTTTVKF NRRKVMDSDE DDDY | |
Greater than 80% as determined by SDS-PAGE. |
9141 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |