missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMA5 Protein
A cDNA sequence encoding the PSMA5 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13184-1ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
PSMA5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMFLT RSEYDRGVNT FSPEGRLFQV EYAIEAIKLG STAIGIQTSE GVCLAVEKRI TSPLMEPSSI EKIVEIDAHI GCAMSGLIAD AKTLIDKARV ETQNHWFTYN ETMTVESVTQ AVSNLALQFG EEDADPGAMS RPFGVALLFG GVDEKGPQLF HMDPSGTFVQ CDARAIGSAS EGAQSSLQEV YHKSMTLKEA IKSSLIILKQ VMEEKLNATN IELATVQPGQ NFHMFTKEEL EEVIKDI | |
Greater than 90% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PSMA5 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 100mM NaCl. |