missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RASSF1A Protein
A cDNA sequence encoding the RASSF1A was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13273-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
RASSF1A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMSGE PELIELRELA PAGRAGKGRT RLERANALRI ARGTACNPTR QLVPGRGHRF QPAGPATHTW CDLCGDFIWG VVRKGLQCAH CKFTCHYRCR ALVCLDCCGP RDLGWEPAVE RDTNVDEPVE WETPDLSQAE IEQKIKEYNA QINSNLFMSL NKDGSYTGFI KVQLKLVRPV SVPSSKKPPS LQDARRGPGR GTSVRRRTSF YLPKDAVKHL HVLSRTRARE VIEALLRKFL VVDDPRKFAL FERAERHGQV YLRKLLDDEQ PLRLRLLAGP SDKALSFVLK ENDSGEVNWD AFSMPELHNF LRILQREEEE HLRQILQKYS YCRQKIQEAL HACPLG | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RASSF1A protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) 0.4M Urea and 10% glycerol. |