missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SDHAF1 Protein
A cDNA sequence encoding the SDHAF1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13441-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
644096 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSRHSRL QRQVLSLYRD LLRAGRGKPG AEARVRAEFR QHAGLPRSDV LRIEYLYRRG RRQLQLLRSG HATAMGAFVR PRAPTGEPGG VGSQPDDGDS PRNPHDSTGA PETRPDGR | |
Greater than 85.0% as determined by SDS-PAGE. |
SDHAF1 Protein | |
Research Use Only | |
SDHAF1 | |
Human | |
His | |
SDHAF1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 30% glycerol, 2mM DTT and 0.1mM PMSF. |