missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNNI3 Protein
A cDNA sequence encoding the TNNI3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13777-HIS-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
TNNI3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MADGSSDAAR EPRPAPAPIR RRSSNYRAYA TEPHAKKKSK ISASRKLQLK TLLLQIAKQE LEREAEERRG EKGRALSTRC QPLELAGLGF AELQDLCRQL HARVDKVDEE RYDIEAKVTK NITEIADLTQ KIFDLRGKFK RPTLRRVRIS ADAMMQALLG ARAKESLDLR AHLKQVKKED TEKENREVGD WRKNIDALSG MEGRKKKFES | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TNNI3 protein solution in PBS and 25mM arginine. |