missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2L6 Protein
A cDNA sequence encoding the UBE2L6 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13864-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
UBE2L6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMASM RVVKELEDLQ KKPPPYLRNL SSDDANVLVW HALLLPDQPP YHLKAFNLRI SFPPEYPFKP PMIKFTTKIY HPNVDENGQI CLPIISSENW KPCTKTCQVL EALNVLVNRP NIREPLRMDL ADLLTQNPEL FRKNAEEFTL RFGVDRPS | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein contains 20mM Tris pH 8.0, 0.1mM PMSF, 1mM DTT, and 10% glycerol. |